Anti-PEX5 Mouse Monoclonal Antibody [clone: 6E9]

Proveedor: OriGene
CF501430
ORIGCF501430EA 764 EUR
ORIGCF501430
Anti-PEX5 Mouse Monoclonal Antibody [clone: 6E9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PEX5
Clonality: Monoclonal
Clone: 6E9
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR