Anti-PANK2 Mouse Monoclonal Antibody [clone: 3G4]

Proveedor: OriGene
CF501419
ORIGCF501419EA 764 EUR
ORIGCF501419
Anti-PANK2 Mouse Monoclonal Antibody [clone: 3G4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PANK2
Clonality: Monoclonal
Clone: 3G4
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR