Anti-KHK Mouse Monoclonal Antibody [clone: 3D1]

Proveedor: OriGene
ORIGCF501410EA 764 EUR
ORIGCF501410
Anti-KHK Mouse Monoclonal Antibody [clone: 3D1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: KHK
Clonality: Monoclonal
Clone: 3D1
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR