Anti-GBE1 Mouse Monoclonal Antibody [clone: 4E6]

Proveedor: OriGene
ORIGCF500779EA 764 EUR
ORIGCF500779
Anti-GBE1 Mouse Monoclonal Antibody [clone: 4E6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: GBE1
Clonality: Monoclonal
Clone: 4E6
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR