Anti-HSPA1A Mouse Monoclonal Antibody [clone: 3G9]

Proveedor: OriGene
CF500771
ORIGCF500771EA 764 EUR
ORIGCF500771
Anti-HSPA1A Mouse Monoclonal Antibody [clone: 3G9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: HSPA1A
Clonality: Monoclonal
Clone: 3G9
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR