Anti-RPA2 Mouse Monoclonal Antibody [clone: 4C3]

Proveedor: OriGene
CF500766
ORIGCF500766EA 764 EUR
ORIGCF500766
Anti-RPA2 Mouse Monoclonal Antibody [clone: 4C3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: RPA2
Clonality: Monoclonal
Clone: 4C3
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR