Anti-RPA2 Mouse Monoclonal Antibody [clone: 10G1]

Proveedor: OriGene
CF500762
ORIGCF500762EA 764 EUR
ORIGCF500762
Anti-RPA2 Mouse Monoclonal Antibody [clone: 10G1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:50; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: RPA2
Clonality: Monoclonal
Clone: 10G1
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR