Anti-HDAC10 Mouse Monoclonal Antibody [clone: 3C2]

Proveedor: OriGene
CF500759
ORIGCF500759EA 786 EUR
ORIGCF500759
Anti-HDAC10 Mouse Monoclonal Antibody [clone: 3C2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50 Flow Cytometry: 1:100

Type: Primary
Antigen: HDAC10
Clonality: Monoclonal
Clone: 3C2
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR