Anti-PRKCE Mouse Monoclonal Antibody [clone: 1B7]

Proveedor: OriGene
CF502395
ORIGCF502395EA 764 EUR
ORIGCF502395
Anti-PRKCE Mouse Monoclonal Antibody [clone: 1B7]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PRKCE
Clonality: Monoclonal
Clone: 1B7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR