Anti-VCAM1 Mouse Monoclonal Antibody [clone: 3H10]

Proveedor: OriGene
CF502391
ORIGCF502391EA 764 EUR
ORIGCF502391
Anti-VCAM1 Mouse Monoclonal Antibody [clone: 3H10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: VCAM1
Clonality: Monoclonal
Clone: 3H10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR