Anti-PLK1 Mouse Monoclonal Antibody [clone: 6C6]

Proveedor: OriGene
ORIGCF500401EA 764 EUR
ORIGCF500401
Anti-PLK1 Mouse Monoclonal Antibody [clone: 6C6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:150; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: PLK1
Clonality: Monoclonal
Clone: 6C6
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR