Anti-EIF2S1 Mouse Monoclonal Antibody [clone: 5D10]

Proveedor: OriGene
CF501401
ORIGCF501401EA 764 EUR
ORIGCF501401
Anti-EIF2S1 Mouse Monoclonal Antibody [clone: 5D10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: EIF2S1
Clonality: Monoclonal
Clone: 5D10
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR