Anti-GORASP1 Mouse Monoclonal Antibody [clone: 4F8]

Proveedor: OriGene
CF501392
ORIGCF501392EA 764 EUR
ORIGCF501392
Anti-GORASP1 Mouse Monoclonal Antibody [clone: 4F8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: GORASP1
Clonality: Monoclonal
Clone: 4F8
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR