Anti-COX6B2 Mouse Monoclonal Antibody [clone: 1F5]

Proveedor: OriGene
CF501377
ORIGCF501377EA 764 EUR
ORIGCF501377
Anti-COX6B2 Mouse Monoclonal Antibody [clone: 1F5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100

Type: Primary
Antigen: COX6B2
Clonality: Monoclonal
Clone: 1F5
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR