Anti-PRL Mouse Monoclonal Antibody [clone: 2B7]

Proveedor: OriGene
CF500721
ORIGCF500721EA 786 EUR
ORIGCF500721
Anti-PRL Mouse Monoclonal Antibody [clone: 2B7]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: PRL
Clonality: Monoclonal
Clone: 2B7
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR