Anti-PDE2A Mouse Monoclonal Antibody [clone: 2F1]

Proveedor: OriGene
ORIGCF502354EA 764 EUR
ORIGCF502354
Anti-PDE2A Mouse Monoclonal Antibody [clone: 2F1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: PDE2A
Clonality: Monoclonal
Clone: 2F1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR