Anti-CTNNB1 Mouse Monoclonal Antibody [clone: 9H2]

Proveedor: OriGene
CF502341
ORIGCF502341EA 764 EUR
ORIGCF502341
Anti-CTNNB1 Mouse Monoclonal Antibody [clone: 9H2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: CTNNB1
Clonality: Monoclonal
Clone: 9H2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR