Anti-SERPINA1 Mouse Monoclonal Antibody [clone: 8E3]

Proveedor: OriGene
CF500376
ORIGCF500376EA 764 EUR
ORIGCF500376
Anti-SERPINA1 Mouse Monoclonal Antibody [clone: 8E3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:150; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: SERPINA1
Clonality: Monoclonal
Clone: 8E3
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR