Anti-NEUROG1 Mouse Monoclonal Antibody [clone: 10B10]

Proveedor: OriGene
CF500354
ORIGCF500354EA 764 EUR
ORIGCF500354
Anti-NEUROG1 Mouse Monoclonal Antibody [clone: 10B10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1000; Immunoprecipitation: 2-4?g/mg

Type: Primary
Antigen: NEUROG1
Clonality: Monoclonal
Clone: 10B10
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR