Anti-L1CAM Mouse Monoclonal Antibody [clone: 1H3]

Proveedor: OriGene
CF500353
ORIGCF500353EA 764 EUR
ORIGCF500353
Anti-L1CAM Mouse Monoclonal Antibody [clone: 1H3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2500-5000; Immunofluorescence: 1:100; Flow Cytometry: 1:100; Immunoprecipitation: 2-4?g/mg

Type: Primary
Antigen: L1CAM
Clonality: Monoclonal
Clone: 1H3
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR