Anti-PROM2 Mouse Monoclonal Antibody [clone: 3D12]

Proveedor: OriGene
CF500349
ORIGCF500349EA 786 EUR
ORIGCF500349
Anti-PROM2 Mouse Monoclonal Antibody [clone: 3D12]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: PROM2
Clonality: Monoclonal
Clone: 3D12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR