Anti-GFAP Mouse Monoclonal Antibody [clone: 2D8]

Proveedor: OriGene
CF500341
ORIGCF500341EA 786 EUR
ORIGCF500341
Anti-GFAP Mouse Monoclonal Antibody [clone: 2D8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000-2000; Immunofluorescence: 1:50-100

Type: Primary
Antigen: GFAP
Clonality: Monoclonal
Clone: 2D8
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR