Anti-C17orf28 Mouse Monoclonal Antibody [clone: 2G6]

Proveedor: OriGene
CF501371
ORIGCF501371EA 764 EUR
ORIGCF501371
Anti-C17orf28 Mouse Monoclonal Antibody [clone: 2G6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: C17orf28
Clonality: Monoclonal
Clone: 2G6
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR