Anti-C17orf28 Mouse Monoclonal Antibody [clone: 3B5]

Proveedor: OriGene
CF501359
ORIGCF501359EA 764 EUR
ORIGCF501359
Anti-C17orf28 Mouse Monoclonal Antibody [clone: 3B5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: C17orf28
Clonality: Monoclonal
Clone: 3B5
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR