Anti-TBXAS1 Mouse Monoclonal Antibody [clone: 2C1]

Proveedor: OriGene
CF501358
ORIGCF501358EA 764 EUR
ORIGCF501358
Anti-TBXAS1 Mouse Monoclonal Antibody [clone: 2C1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: TBXAS1
Clonality: Monoclonal
Clone: 2C1
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR