Anti-SNX9 Mouse Monoclonal Antibody [clone: 2G4]

Proveedor: OriGene
CF501356
ORIGCF501356EA 764 EUR
ORIGCF501356
Anti-SNX9 Mouse Monoclonal Antibody [clone: 2G4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SNX9
Clonality: Monoclonal
Clone: 2G4
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR