Anti-MOBKL1A Mouse Monoclonal Antibody [clone: 1H8]

Proveedor: OriGene
CF501351
ORIGCF501351EA 764 EUR
ORIGCF501351
Anti-MOBKL1A Mouse Monoclonal Antibody [clone: 1H8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: MOBKL1A
Clonality: Monoclonal
Clone: 1H8
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR