Anti-TTC32 Mouse Monoclonal Antibody [clone: 1F7]

Proveedor: OriGene
CF501337
ORIGCF501337EA 786 EUR
ORIGCF501337
Anti-TTC32 Mouse Monoclonal Antibody [clone: 1F7]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: TTC32
Clonality: Monoclonal
Clone: 1F7
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR