Anti-CD5 Mouse Monoclonal Antibody [clone: 7A7]

Proveedor: OriGene
CF501335
ORIGCF501335EA 764 EUR
ORIGCF501335
Anti-CD5 Mouse Monoclonal Antibody [clone: 7A7]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: 7A7
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR