Anti-PSMA7 Mouse Monoclonal Antibody [clone: 8F9]

Proveedor: OriGene
ORIGCF500694EA 764 EUR
ORIGCF500694
Anti-PSMA7 Mouse Monoclonal Antibody [clone: 8F9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: PSMA7
Clonality: Monoclonal
Clone: 8F9
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR