Anti-NUDT6 Mouse Monoclonal Antibody [clone: 6H3]

Proveedor: OriGene
ORIGCF502334EA 764 EUR
ORIGCF502334
Anti-NUDT6 Mouse Monoclonal Antibody [clone: 6H3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: NUDT6
Clonality: Monoclonal
Clone: 6H3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR