Anti-GFAP Mouse Monoclonal Antibody [clone: 5D7]

Proveedor: OriGene
CF500340
ORIGCF500340EA 786 EUR
ORIGCF500340
Anti-GFAP Mouse Monoclonal Antibody [clone: 5D7]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50

Type: Primary
Antigen: GFAP
Clonality: Monoclonal
Clone: 5D7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR