Anti-NEUROG1 Mouse Monoclonal Antibody [clone: 15A7]

Proveedor: OriGene
CF500303
ORIGCF500303EA 764 EUR
ORIGCF500303
Anti-NEUROG1 Mouse Monoclonal Antibody [clone: 15A7]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Immunoprecipitation: 2-4?g/mg

Type: Primary
Antigen: NEUROG1
Clonality: Monoclonal
Clone: 15A7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR