Anti-TNNI3 Mouse Monoclonal Antibody [clone: 4A3]

Proveedor: OriGene
CF500256
ORIGCF500256EA 764 EUR
ORIGCF500256
Anti-TNNI3 Mouse Monoclonal Antibody [clone: 4A3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50

Type: Primary
Antigen: TNNI3
Clonality: Monoclonal
Clone: 4A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR