Anti-C9orf41 Mouse Monoclonal Antibody [clone: 6E4]

Proveedor: OriGene
CF501300
ORIGCF501300EA 764 EUR
ORIGCF501300
Anti-C9orf41 Mouse Monoclonal Antibody [clone: 6E4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: C9orf41
Clonality: Monoclonal
Clone: 6E4
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR