Anti-C9orf41 Mouse Monoclonal Antibody [clone: 1E8]

Proveedor: OriGene
CF501299
ORIGCF501299EA 764 EUR
ORIGCF501299
Anti-C9orf41 Mouse Monoclonal Antibody [clone: 1E8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: C9orf41
Clonality: Monoclonal
Clone: 1E8
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR