Anti-FCGR2A Mouse Monoclonal Antibody [clone: 6D11]

Proveedor: OriGene
CF500653
ORIGCF500653EA 764 EUR
ORIGCF500653
Anti-FCGR2A Mouse Monoclonal Antibody [clone: 6D11]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-1:1000; Immunochemistry: 1:50; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: FCGR2A
Clonality: Monoclonal
Clone: 6D11
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR