Anti-FCGR2A Mouse Monoclonal Antibody [clone: 13G9]

Proveedor: OriGene
CF500646
ORIGCF500646EA 764 EUR
ORIGCF500646
Anti-FCGR2A Mouse Monoclonal Antibody [clone: 13G9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: FCGR2A
Clonality: Monoclonal
Clone: 13G9
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR