Anti-GBP2 Mouse Monoclonal Antibody [clone: 5C8]

Proveedor: OriGene
CF500640
ORIGCF500640EA 786 EUR
ORIGCF500640
Anti-GBP2 Mouse Monoclonal Antibody [clone: 5C8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: GBP2
Clonality: Monoclonal
Clone: 5C8
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR