Anti-PDE10A Mouse Monoclonal Antibody [clone: 3G9]

Proveedor: OriGene
CF500638
ORIGCF500638EA 764 EUR
ORIGCF500638
Anti-PDE10A Mouse Monoclonal Antibody [clone: 3G9]
Anticuerpos
Recommended Dilutions: Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PDE10A
Clonality: Monoclonal
Clone: 3G9
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR