Anti-PCSK7 Mouse Monoclonal Antibody [clone: 1B8]

Proveedor: OriGene
CF500628
ORIGCF500628EA 764 EUR
ORIGCF500628
Anti-PCSK7 Mouse Monoclonal Antibody [clone: 1B8]
Anticuerpos
Recommended Dilutions: Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PCSK7
Clonality: Monoclonal
Clone: 1B8
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR