Anti-ACY3 Mouse Monoclonal Antibody [clone: 2D5]

Proveedor: OriGene
CF502295
ORIGCF502295EA 764 EUR
ORIGCF502295
Anti-ACY3 Mouse Monoclonal Antibody [clone: 2D5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ACY3
Clonality: Monoclonal
Clone: 2D5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR