Anti-ACY3 Mouse Monoclonal Antibody [clone: 2G4]

Proveedor: OriGene
CF502290
ORIGCF502290EA 764 EUR
ORIGCF502290
Anti-ACY3 Mouse Monoclonal Antibody [clone: 2G4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Flow Cytometry: 1:100

Type: Primary
Antigen: ACY3
Clonality: Monoclonal
Clone: 2G4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR