Anti-PIP4K2A Mouse Monoclonal Antibody [clone: 1B2]

Proveedor: OriGene
CF502285
ORIGCF502285EA 764 EUR
ORIGCF502285
Anti-PIP4K2A Mouse Monoclonal Antibody [clone: 1B2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PIP4K2A
Clonality: Monoclonal
Clone: 1B2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR