Anti-TNNI3 Mouse Monoclonal Antibody [clone: 3G2]

Proveedor: OriGene
ORIGCF500255EA 764 EUR
ORIGCF500255
Anti-TNNI3 Mouse Monoclonal Antibody [clone: 3G2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50

Type: Primary
Antigen: TNNI3
Clonality: Monoclonal
Clone: 3G2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR