Anti-C-Jun Mouse Monoclonal Antibody [clone: 1C8]

Proveedor: OriGene
CF500226
ORIGCF500226EA 764 EUR
ORIGCF500226
Anti-C-Jun Mouse Monoclonal Antibody [clone: 1C8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: C-Jun
Clonality: Monoclonal
Clone: 1C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR