Anti-IL3 Mouse Monoclonal Antibody [clone: 5G7H9]

Proveedor: OriGene
ORIGCF500204EA 764 EUR
ORIGCF500204
Anti-IL3 Mouse Monoclonal Antibody [clone: 5G7H9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: IL3
Clonality: Monoclonal
Clone: 5G7H9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR