Anti-HHex Mouse Monoclonal Antibody [clone: 3F4 1B1]

Proveedor: OriGene
CF500099
ORIGCF500099EA 764 EUR
ORIGCF500099
Anti-HHex Mouse Monoclonal Antibody [clone: 3F4 1B1]
Anticuerpos
Recommended Dilutions: Western Blot: 110000; Immunofluorescence: 1:100

Type: Primary
Antigen: HHex
Clonality: Monoclonal
Clone: 3F4 1B1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR