Anti-PPP1R7 Mouse Monoclonal Antibody [clone: 4F9]

Proveedor: OriGene
CF500614
ORIGCF500614EA 786 EUR
ORIGCF500614
Anti-PPP1R7 Mouse Monoclonal Antibody [clone: 4F9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:50-100

Type: Primary
Antigen: PPP1R7
Clonality: Monoclonal
Clone: 4F9
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR