Anti-POR Mouse Monoclonal Antibody [clone: 5B6]

Proveedor: OriGene
ORIGCF500613EA 764 EUR
ORIGCF500613
Anti-POR Mouse Monoclonal Antibody [clone: 5B6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: POR
Clonality: Monoclonal
Clone: 5B6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR